Learn More
Indiana Fluid Systems Technologies SS FF QK CON STEM W/OUT VLV
Catalog No. NC2104110
Encompass
Stainless Steel Full Flow Quick Connect Stem without Valve, 11.5 Cv, 1/2 in. Female NPT |   BODY MATERIAL: 316 Stainless Steel 1/2 in. |CONNECTION 1 SIZE: 1/2 in. |CONNECTION 1 TYPE: Female NPT |O-RING MATERIAL: Fluorocarbon FKM QF8 |SERIES: QF8
This item is not returnable. View return policy
Catalog No. NC2104110 Supplier Indiana Fluid Systems Technologies Supplier No. SSQF8S8PF
Stainless Steel Full Flow Quick Connect Stem without Valve, 11.5 Cv, 1/2 in. Female NPT |   BODY MATERIAL: 316 Stainless Steel 1/2 in. |CONNECTION 1 SIZE: 1/2 in. |CONNECTION 1 TYPE: Female NPT |O-RING MATERIAL: Fluorocarbon FKM QF8 |SERIES: QF8