Learn More
Uline Stainless Steel Filler Table - 18 x 30, Stainless Steel
Catalog No. NC2291459
Encompass
All stainless steel construction Type 304 top. Narrow footprint for squeezing into tight spaces. 35 table height. Generous 4 high backsplash. 1 5/8 diameter legs with adjustable bottom shelf and leveling feet. NSF certified. Optional Casters add mobility.
Catalog No. NC2291459 Supplier Uline Supplier No. H9747
All stainless steel construction Type 304 top. Narrow footprint for squeezing into tight spaces. 35 table height. Generous 4 high backsplash. 1 5/8 diameter legs with adjustable bottom shelf and leveling feet. NSF certified. Optional Casters add mobility.