Learn More
Cole-Parmer Stainless-steel Griffin-style beaker with easy-pour rim, 250 mL
Catalog No. NC0394632 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Stainless-steel Griffin-style beaker with easy-pour rim, 250 mLGet durable, economical mixing capabilitiesSturdy beakers for general stirring or measuring of solutionsBeaker features a 250 mL capacityThese Griffin-style beakers with pouring spout have full capacity mark near the top to assure accurate measuring. Constructed of medium gauge 304 stainless steel, these beakers are durable and easy to clean.
Catalog No. NC0394632 Supplier Cole-Parmer Supplier No. 0720540
Cole-Parmer Stainless-steel Griffin-style beaker with easy-pour rim, 250 mLGet durable, economical mixing capabilitiesSturdy beakers for general stirring or measuring of solutionsBeaker features a 250 mL capacityThese Griffin-style beakers with pouring spout have full capacity mark near the top to assure accurate measuring. Constructed of medium gauge 304 stainless steel, these beakers are durable and easy to clean.