Learn More
Analytical Sales & Services SS High Flow 1.0mL Ternary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000804227
Encompass
Advantage Stainless Steel High Flow 1.0mL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-04227 Supplier Analytical Sales & Services Supplier No. 133011100
Advantage Stainless Steel High Flow 1.0mL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH