Learn More
Analytical Sales & Services SS High Flow 1.5mL Inline Static Bed Mixer - Complete Assembly
Catalog No. 5000807037
Encompass
Advantage Stainless Steel High Flow 1.5mL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-07037 Supplier Analytical Sales & Services Supplier No. 130015000
Advantage Stainless Steel High Flow 1.5mL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH