Learn More
Analytical Sales & Services SS High Flow 800µL Binary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000804452
Encompass
Advantage Stainless Steel High Flow 800µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-04452 Supplier Analytical Sales & Services Supplier No. 132080000
Advantage Stainless Steel High Flow 800µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH