Learn More
Analytical Sales & Services SS High Flow 800µL Ternary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000806598
Encompass
Advantage Stainless Steel High Flow 800µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-06598 Supplier Analytical Sales & Services Supplier No. 133080000
Advantage Stainless Steel High Flow 800µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH