Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Hose Clamps, Stainless Steel, 1-1/16""ID x 2"" ID; 10/Box
Catalog No. NC1279557
Encompass
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.
Catalog No. NC1279557 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640314
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.