Learn More
New Jersey Fluid Systems LLC SS HOSE CONNECT 1/4IN STF 1/4
Catalog No. NC2070997
Encompass
Stainless Steel Hose Connector, 1/4 in. Swagelok Tube Fitting, 1/4 in. Hose ID
Catalog No. NC2070997 Supplier New Jersey Fluid Systems LLC Supplier No. SS4HC1400
Stainless Steel Hose Connector, 1/4 in. Swagelok Tube Fitting, 1/4 in. Hose ID