Learn More
Hach Company SS Insert. Mt. Hardware-2in
Catalog No. 5000207081 Shop All Hach Company Products
Encompass
Insertion Mounting Hardware-2 -Stainless Steel-3700 SeriesStainless steel 2in NPT insertion assembly with ball valve For use with the 3700-series inductive/electrodeless conductivity sensors
Catalog No. 50-002-07081 Supplier Hach Company Supplier No. MH118M9NZ
Add to Cart
Add to Cart
Request a Quote
Insertion Mounting Hardware-2 -Stainless Steel-3700 SeriesStainless steel 2in NPT insertion assembly with ball valve For use with the 3700-series inductive/electrodeless conductivity sensors