Learn More
Hach Company SS Insertion Mounting Hardware
Catalog No. 5000207150 Shop All Hach Company Products
Encompass
The insertion hardware includes a 1 in ball valve 1 in NPT close nipple for process connection sensor connection tube stainless steel extension pipe and stainless steel compression fitting with washer and lock nut
Catalog No. 50-002-07150 Supplier Hach Company Supplier No. MH116M3MZ
The insertion hardware includes a 1 in ball valve 1 in NPT close nipple for process connection sensor connection tube stainless steel extension pipe and stainless steel compression fitting with washer and lock nut