Learn More
Med Vet International Stainless Steel Instrument Tray, 10" x 6-1/2" x 3/4"
Supplier:  Med Vet International 49365
Encompass_Preferred
Stainless Steel Hospital Ware is manufactured to the exacting standards of the healthcare industry. Every product is manufactured using seamless technology which eliminates areas that can harbor bacteria growth. All utensils are manufactured of 18-8 nickel-bearing stainless steel to prevent rust, and polished to a mirror finish.
Catalog No. 50-223-2261