Learn More
Analytical Sales & Services SS Low Flow 150µL Binary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000807036
Encompass
Advantage Stainless Low Flow 150µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-07036 Supplier Analytical Sales & Services Supplier No. 130011501
Advantage Stainless Low Flow 150µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH