Learn More
Analytical Sales & Services SS Low Flow 250µL Inline Static Bed Mixer - Complete Assembly
Catalog No. 5000807273
Encompass
Advantage Stainless Steel Low Flow 250µL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-07273 Supplier Analytical Sales & Services Supplier No. 130022501
Advantage Stainless Steel Low Flow 250µL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH