Learn More
Analytical Sales & Services SS Low Flow 50µL Ternary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000806021
Encompass
Advantage Stainless Steel Low Flow 50µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-06021 Supplier Analytical Sales & Services Supplier No. 130030501
Advantage Stainless Steel Low Flow 50µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH