Learn More
Electron Microscopy Sciences Stainless Steel Matrices Rat Heart
SDP
Catalog No. 5033902
Encompass_Preferred
Stainless steel matrices with 0.5 mm channel for slicing rodent brain, heart, and similar organs as thin as possible. Made of high-quality stainless steel. They can be repeatedly used, cleaned, autoclaved, and frozen. Produce fine slices. Accurate sectioning. Easy cleaning and easy handling.
Catalog No. 50-339-02 Supplier Electron Microscopy Sciences Supplier No. 69096H
Stainless steel matrices with 0.5 mm channel for slicing rodent brain, heart, and similar organs as thin as possible. Made of high-quality stainless steel. They can be repeatedly used, cleaned, autoclaved, and frozen. Produce fine slices. Accurate sectioning. Easy cleaning and easy handling.