Learn More
Electron Microscopy Sciences Stainless Steel Matrices Small Rat, Sagital, 175-300g
SDP
Catalog No. 5033904
Encompass_Preferred
Stainless steel matrices with 0.5 mm channel for slicing rodent brain, heart, and similar organs as thin as possible. Made of high-quality stainless steel. They can be repeatedly used, cleaned, autoclaved, and frozen. Produce fine slices. Accurate sectioning. Easy cleaning and easy handling.
Type: Sagital
Size: 175-300g
Catalog No. 50-339-04 Supplier Electron Microscopy Sciences Supplier No. 69091S
Add to Cart
Add to Cart
Stainless steel matrices with 0.5 mm channel for slicing rodent brain, heart, and similar organs as thin as possible. Made of high-quality stainless steel. They can be repeatedly used, cleaned, autoclaved, and frozen. Produce fine slices. Accurate sectioning. Easy cleaning and easy handling.
Type: Sagital
Size: 175-300g