Learn More
Analytical Sales & Services SS Micro Flow 0.5µL Binary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000806568
Encompass
Advantage Stainless Steel Micro Flow 0.5µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-06568 Supplier Analytical Sales & Services Supplier No. 130024031
Advantage Stainless Steel Micro Flow 0.5µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH