Learn More
Analytical Sales & Services SS Micro Flow 0.5µL Inline Static Bed Mixer - Complete Assembly
Catalog No. 5000806957
Encompass
Advantage Stainless Steel Micro Flow 0.5µL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-06957 Supplier Analytical Sales & Services Supplier No. 130020031
Advantage Stainless Steel Micro Flow 0.5µL Inline Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH