Learn More
Analytical Sales & Services SS Micro Flow 10µL Binary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000806567
Encompass
Advantage Stainless Steel Micro Flow 10µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-06567 Supplier Analytical Sales & Services Supplier No. 130024010
Add to Cart
Add to Cart
Advantage Stainless Steel Micro Flow 10µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH