Learn More
Analytical Sales & Services SS Micro Flow 2.0µL Binary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000807089
Encompass
Advantage Stainless Steel Micro Flow 2.0µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-07089 Supplier Analytical Sales & Services Supplier No. 130024002
Advantage Stainless Steel Micro Flow 2.0µL Binary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH