Learn More
Analytical Sales & Services SS Micro Flow 5.0µL Static Bed Mixer - Cartridge Only
Catalog No. 5000807088
Encompass
Advantage Stainless Steel Micro Flow 5.0µL Static Bed Mixer Cartridge Only
Qty/Pkg: EACH
Catalog No. 50-008-07088 Supplier Analytical Sales & Services Supplier No. 130000051
Advantage Stainless Steel Micro Flow 5.0µL Static Bed Mixer Cartridge Only
Qty/Pkg: EACH