Learn More
Analytical Sales & Services SS Micro Flow 5.0µL Ternary Tee Static Bed Mixer - Complete Assembly
Catalog No. 5000804284
Encompass
Advantage Stainless Steel Micro Flow 5.0µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH
Catalog No. 50-008-04284 Supplier Analytical Sales & Services Supplier No. 130034005
Advantage Stainless Steel Micro Flow 5.0µL Ternary Tee Static Bed Mixer Complete Assembly Includes Cartridge and Housing
Qty/Pkg: EACH