Learn More
Analytical Sales & Services SS Micro Flow Binary Static Bed Mixer - Housing Only
Catalog No. 5000807035
Encompass
Advantage Stainless Steel Micro Flow Binary Tee Static Bed Mixer Housing Only\n\nQty/Pkg: EACH
Catalog No. 50-008-07035 Supplier Analytical Sales & Services Supplier No. 130024000
Add to Cart
Add to Cart
Advantage Stainless Steel Micro Flow Binary Tee Static Bed Mixer Housing Only\n\nQty/Pkg: EACH