Learn More
Inter Dyne Systems Inc Stainless Steel Pegboard 24x24 inch
Catalog No. NC1223004
Encompass_Preferred
The Victoria pegboard is constructed from 20 gauge, type 304 stainless steel, polished to a #4 finish. The V style pegboard comes in 14 standard sizes and a variety of mounting options. Victoria pegboards are designed with an integral drip trough. All boards come standard with a wall bracket, standard 6 white pegs and 3 ft. of clear plastic drain tubing.
Catalog No. NC1223004 Supplier Inter Dyne Systems Inc Supplier No. V2424
The Victoria pegboard is constructed from 20 gauge, type 304 stainless steel, polished to a #4 finish. The V style pegboard comes in 14 standard sizes and a variety of mounting options. Victoria pegboards are designed with an integral drip trough. All boards come standard with a wall bracket, standard 6 white pegs and 3 ft. of clear plastic drain tubing.