Learn More
SWAGELOCK SS PIPE FITTING, REDUCING ADAP
Catalog No. NC2046215
Encompass
stainless steel Pipe Fitting, Reducing Adapter, 1/4 in. Female NPT x 1/8 in. Male NPT
Catalog No. NC2046215 Supplier SWAGELOCK Supplier No. SS4RA2
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
stainless steel Pipe Fitting, Reducing Adapter, 1/4 in. Female NPT x 1/8 in. Male NPT