Learn More
MSC Vikan Tube Brush, Medium, Red, Polyester, Stainless Steel, 3.5 in Diameter
Catalog No. 502839428
Encompass
This versatile tube brush is designed for cleaning pipes found in dairies, breweries, and the fishing industry. It is suitable for a wide range of food contact applications and can be used with compatible flexible and extension handles.
  • Purpose-built for hygiene and food safety in food manufacturing, retail, and service.
  • Medium bristles effectively scrub with cleaning solutions.
  • Designed for effective pipe, tube, and valve cleaning.
  • Durable construction provides long-lasting performance.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Effective on stubborn debris and residue.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain for hygiene control.
  • Bristles are twisted directly into the brush, reducing the risk of bristle loss.
Catalog No. 50-283-9428 Supplier MSC Supplier No. 65429664
This versatile tube brush is designed for cleaning pipes found in dairies, breweries, and the fishing industry. It is suitable for a wide range of food contact applications and can be used with compatible flexible and extension handles.
  • Purpose-built for hygiene and food safety in food manufacturing, retail, and service.
  • Medium bristles effectively scrub with cleaning solutions.
  • Designed for effective pipe, tube, and valve cleaning.
  • Durable construction provides long-lasting performance.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Effective on stubborn debris and residue.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain for hygiene control.
  • Bristles are twisted directly into the brush, reducing the risk of bristle loss.