Learn More
MSC ASI-American Specialties Paper Towel Dispenser, Surface Mounted, 1 ea
Catalog No. 502818236
Encompass
This surface-mounted unit combines a paper towel dispenser with a waste receptacle, constructed from durable 304 stainless steel. It is designed for efficient and hygienic towel dispensing and waste management in various environments.
  • Dispenses 600 C-fold or 800 multi-fold paper towels.
  • Features a 12 gal (46 L) stainless steel waste receptacle.
  • Equipped with tumbler locks for security and unauthorized access prevention.
  • Heavy-duty, staked piano hinges ensure durability and a neat appearance.
  • Hemmed opening provides safe and easy towel dispensing.
  • Constructed from 22 gauge Type 304 stainless steel with a satin finish.
  • Designed for surface mounting using concealed screws.
  • Self-feeding towel mechanism allows partial load reloading.
Catalog No. 50-281-8236 Supplier MSC Supplier No. 11774478
This surface-mounted unit combines a paper towel dispenser with a waste receptacle, constructed from durable 304 stainless steel. It is designed for efficient and hygienic towel dispensing and waste management in various environments.
  • Dispenses 600 C-fold or 800 multi-fold paper towels.
  • Features a 12 gal (46 L) stainless steel waste receptacle.
  • Equipped with tumbler locks for security and unauthorized access prevention.
  • Heavy-duty, staked piano hinges ensure durability and a neat appearance.
  • Hemmed opening provides safe and easy towel dispensing.
  • Constructed from 22 gauge Type 304 stainless steel with a satin finish.
  • Designed for surface mounting using concealed screws.
  • Self-feeding towel mechanism allows partial load reloading.