Learn More
Analytical Sales & Services SS Prep Tubing 1/8" OD - 0.08" ID x 1M
Catalog No. 5000806675
Encompass
Stainless Steel Prep Tubing 1/8" OD - 0.08" ID x 1M
Catalog No. 50-008-06675 Supplier Analytical Sales & Services Supplier No. 64800
Stainless Steel Prep Tubing 1/8" OD - 0.08" ID x 1M