Learn More
Ancare Corporation SS RAISED WIRE FLOORS 10/CS
SDP
Catalog No. NC1626551
Encompass
Mice. Compatible with our N10 cage. 3 openings per inch.
Catalog No. NC1626551 Supplier Ancare Corporation Supplier No. N10SSRWF
Mice. Compatible with our N10 cage. 3 openings per inch.