Learn More
Invivogen SS RNA40 (LYOPHILIZED ) 100MG
SDP
Catalog No. NC9207760
Encompass
Lyophilized Single Stranded RNA40/ LyoVec, Working Concentration: 0.1 to 10microgram/mL, Quantity: 100microgram
Catalog No. NC9207760 Supplier Invivogen Supplier No. TLRLLRNA40
Lyophilized Single Stranded RNA40/ LyoVec, Working Concentration: 0.1 to 10microgram/mL, Quantity: 100microgram