Learn More
Perkin Elmer US LLC Stainless Sample Probe, No Tubing
Catalog No. 509042744
Encompass_Preferred
Sampling Probe with Screw Fitting for PerkinElmer AS-90/90A/90 Plus/91/93 Plus Autosampler, Requires but does not include capillary tubing assembly (P/N B3000157, B3000158, or B0191060).
Catalog No. 50-904-2744 Supplier Perkin Elmer US LLC Supplier No. B3000152
Sampling Probe with Screw Fitting for PerkinElmer AS-90/90A/90 Plus/91/93 Plus Autosampler, Requires but does not include capillary tubing assembly (P/N B3000157, B3000158, or B0191060).