Learn More
Cole-Parmer Stainless steel sanitary bottle; 1 liter, 2" flange
Supplier:  Cole-Parmer 0750601
Encompass
Sanitary bottles are made from sanitary 316L stainless steel with #9, 24 Ra finish on interior and exterior. Bottles feature a sanitary Tri-Clamp flange opening; include an endcap, platinum-cured silicone gasket, and heavy-duty clamp.
Catalog No. NC1659950