Learn More
Sanisure Inc Gamma Stable Stir Bars
Catalog No. NC1947882
Encompass_Preferred
The SaniSure Gamma Stable Stir Bars are the ideal product for simple agitation of biopharmaceutical process solutions. Available standalone or integrated into a variety of container formats, they provide a simple mixing solution and process flexibility. SaniSure Gamma Stable Stir Bars are available in various sizes to suit the desired container volume. They are Animal Derived Components Free, meet USP Class VI, and are lot traceable.
Catalog No. NC1947882 Supplier Sanisure Inc Supplier No. SSSB1080PVDFG
The SaniSure Gamma Stable Stir Bars are the ideal product for simple agitation of biopharmaceutical process solutions. Available standalone or integrated into a variety of container formats, they provide a simple mixing solution and process flexibility. SaniSure Gamma Stable Stir Bars are available in various sizes to suit the desired container volume. They are Animal Derived Components Free, meet USP Class VI, and are lot traceable.