Learn More
Uline Stainless Steel Security Cart
Supplier:  Uline H6841
Encompass
Security cart, stainless steel, 60 x 24 x 69"""", type 304 stainless steel, lockable double doors, 2 heavy duty shelves, 5"""" lockable stainless steel swivel casters
Catalog No. NC2021467