Learn More
Electron Microscopy Sciences Stainless Steel Slide Rack
SDP
Catalog No. 5028709
Encompass_Preferred
A stainless steel slotted rack that holds 50 slides in sizes: 3" x 1", 3" x 1.5 ", and 3" x 2". Rack is resistant to staining solutions and has permanently attached handle.
Catalog No. 50-287-09 Supplier Electron Microscopy Sciences Supplier No. 7031254
A stainless steel slotted rack that holds 50 slides in sizes: 3" x 1", 3" x 1.5 ", and 3" x 2". Rack is resistant to staining solutions and has permanently attached handle.