Learn More
MSC Nilfisk Industrial Vacuum, Electric, HEPA Filter, 26 gal, 1 ea
Catalog No. 502818133
Encompass
The Nilfisk S3 single-phase industrial vacuum is designed for diverse cleaning tasks across industries like food, paper/corrugated, packaging, and woodworking, featuring robust construction and advanced innovations.
  • Modular design for effortless modifications and servicing
  • Electronic keypad with LED indicators to monitor filter performance
  • Manual filter shaker and a standard polyester main filter ensure high filtration efficiency
  • Upstream HEPA filtration retains 99.995% of particles down to 0.18 microns
  • Steel tank material for durability
  • Disposable bag for vacuum collection
  • Includes a 100 L bin and tool tray
Catalog No. 50-281-8133 Supplier MSC Supplier No. 13676002
Add to Cart
Add to Cart
Request a Quote
The Nilfisk S3 single-phase industrial vacuum is designed for diverse cleaning tasks across industries like food, paper/corrugated, packaging, and woodworking, featuring robust construction and advanced innovations.
  • Modular design for effortless modifications and servicing
  • Electronic keypad with LED indicators to monitor filter performance
  • Manual filter shaker and a standard polyester main filter ensure high filtration efficiency
  • Upstream HEPA filtration retains 99.995% of particles down to 0.18 microns
  • Steel tank material for durability
  • Disposable bag for vacuum collection
  • Includes a 100 L bin and tool tray