Learn More
Medicus Health Slide Staining Set, (3) Autoclavable Dishes with Lids, Slide Dipper
SDP
Catalog No. NC3056011
Encompass
Slide Staining Set, (3) Autoclavable Dishes with Lids, (1) Slide Dipper and (1) 12 Place Stainless Steel Dish ∙ Staining station is great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides ∙ Dishes comes complete with detached lids that help reduce reagent evaporation or contamination ∙ Staining Set Overall Dimensions: 9"W x 5"D x 5"H∙ Dish volume is 300 mL ∙ Dish construction withstands repeated autoclaving, even at 150°C.∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides & measures: 4.25"L x 2"W x 4"H
Catalog No. NC3056011 Supplier Medicus Health Supplier No. 7353M1
Slide Staining Set, (3) Autoclavable Dishes with Lids, (1) Slide Dipper and (1) 12 Place Stainless Steel Dish ∙ Staining station is great for immunohistochemistry procedures, histological, cytological and hematological staining procedures as well with total immersion for slides ∙ Dishes comes complete with detached lids that help reduce reagent evaporation or contamination ∙ Staining Set Overall Dimensions: 9"W x 5"D x 5"H∙ Dish volume is 300 mL ∙ Dish construction withstands repeated autoclaving, even at 150°C.∙ Staining Dish is designed for 75mm x 25mm or 3" x 1" slides & measures: 4.25"L x 2"W x 4"H