Learn More
MSC Vikan Stainless Steel Scraper, Threaded Handle, 2, Blue, 1 ea
Catalog No. 502868048
Encompass
This scraper is designed for removing hard-to-reach sticky deposits and dried or burnt-on foods or ingredients. It features a stainless flex steel blade with protective rounded corners, a high-strength attachment, and a threaded handle that fits any color-coded extension handle. It can be used for lighter soils on floors, though not intended for general floor cleaning.
  • Purpose-built for hygiene and food safety in food manufacturing and service
  • Versatile for hand-held or extended handle cleaning
  • Stainless steel resists corrosion and wear
  • Aggressively removes dried or burnt-on materials
  • Lightweight design reduces user fatigue
  • Easy to clean and maintain for hygiene control
  • Color-coded for hygienic zoning plans
Catalog No. 50-286-8048 Supplier MSC Supplier No. 97016323
This scraper is designed for removing hard-to-reach sticky deposits and dried or burnt-on foods or ingredients. It features a stainless flex steel blade with protective rounded corners, a high-strength attachment, and a threaded handle that fits any color-coded extension handle. It can be used for lighter soils on floors, though not intended for general floor cleaning.
  • Purpose-built for hygiene and food safety in food manufacturing and service
  • Versatile for hand-held or extended handle cleaning
  • Stainless steel resists corrosion and wear
  • Aggressively removes dried or burnt-on materials
  • Lightweight design reduces user fatigue
  • Easy to clean and maintain for hygiene control
  • Color-coded for hygienic zoning plans