Learn More
MSC Vikan Scraper, Stainless Steel, 100 mm, Threaded, Red, 1 ea
Catalog No. 502868592
Encompass
This scraper is designed for removing hard-to-reach sticky deposits and dried or burnt-on foods or ingredients. It features a stainless flex steel blade with protective rounded corners, a high-strength attachment, and a threaded handle that is compatible with any extension handle. It can be used for removing lighter soils on floors, though it is not intended for proper floor cleaning.
  • Stainless flex steel blade with protective rounded corners
  • High-strength attachment
  • Threaded handle compatible with extension handles
  • Removes sticky deposits and burnt-on foods
  • Suitable for lighter floor soil removal
Catalog No. 50-286-8592 Supplier MSC Supplier No. 97016257
This scraper is designed for removing hard-to-reach sticky deposits and dried or burnt-on foods or ingredients. It features a stainless flex steel blade with protective rounded corners, a high-strength attachment, and a threaded handle that is compatible with any extension handle. It can be used for removing lighter soils on floors, though it is not intended for proper floor cleaning.
  • Stainless flex steel blade with protective rounded corners
  • High-strength attachment
  • Threaded handle compatible with extension handles
  • Removes sticky deposits and burnt-on foods
  • Suitable for lighter floor soil removal