Learn More
MSC PFERD Block Brush, Straight Back, 6x19 Rows, .012 Stainless Steel Wire, Wooden Block
Catalog No. 502837736
Encompass
This robust block brush is made from wood and designed for heavy-duty cleaning work. It features 302 stainless steel wire, providing excellent corrosion resistance and optimal brush life, suitable for various cleaning applications on stainless steel and other non-ferrous metals.
  • Filament material is stainless steel wire (INOX).
  • Filament material diameter is .012 "
  • Designed for manual use.
  • Features 6x19 rows.
  • Overall length is 7-1/4 "
  • Trim length is 1-3/4 "
  • Straight back version.
  • Width is 2-1/4 "
  • Excellent for cleaning concrete forms.
  • Ergonomic curved handle.
  • Suitable for use on aluminum, bronze, cobalt-based alloys, other non-ferrous metals, and stainless steel (INOX).
  • Applications include cleaning, derusting, removing heat discolouration, removing oxide layers, and roughening.
Catalog No. 50-283-7736 Supplier MSC Supplier No. 49189459
This robust block brush is made from wood and designed for heavy-duty cleaning work. It features 302 stainless steel wire, providing excellent corrosion resistance and optimal brush life, suitable for various cleaning applications on stainless steel and other non-ferrous metals.
  • Filament material is stainless steel wire (INOX).
  • Filament material diameter is .012 "
  • Designed for manual use.
  • Features 6x19 rows.
  • Overall length is 7-1/4 "
  • Trim length is 1-3/4 "
  • Straight back version.
  • Width is 2-1/4 "
  • Excellent for cleaning concrete forms.
  • Ergonomic curved handle.
  • Suitable for use on aluminum, bronze, cobalt-based alloys, other non-ferrous metals, and stainless steel (INOX).
  • Applications include cleaning, derusting, removing heat discolouration, removing oxide layers, and roughening.