Learn More
Ace Glass, Inc. Stainless steel support rod 122cm 48"
SDP
Catalog No. 50977227
Encompass
48 (122cm) long, 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.
Catalog No. 50-977-227 Supplier Ace Glass, Inc. Supplier No. 1117812
48 (122cm) long, 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.