Learn More
Ace Glass, Inc. Stainless steel support rod 244cm 96"
SDP
Catalog No. 50980626
Encompass
96 (244cm) long, 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.
Catalog No. 50-980-626 Supplier Ace Glass, Inc. Supplier No. 1117816
96 (244cm) long, 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.