Learn More
Ace Glass, Inc. Stainless steel support rod 31cm 12"
SDP
Catalog No. 50977224
Encompass
12 (31cm) long 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.
Catalog No. 50-977-224 Supplier Ace Glass, Inc. Supplier No. 1117804
12 (31cm) long 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.