Learn More
MSC Vikan Table and Floor Scraper, Stainless Steel, Stiff, Red, 1 ea
Catalog No. 5000507571
Encompass
This stiff table and floor scraper features a durable, solid stainless steel (AISI 304) blade designed to effectively remove stubborn food debris, residue, and labels from various surfaces. Purpose-built for food manufacturing, commercial kitchens, and food service environments, its robust construction ensures long-lasting performance in hygienic and high-safety applications.
  • Durable, solid stainless steel blade
  • Compatible with all Vikan Euro threaded handles for extended reach
  • Lightweight design to reduce user fatigue
  • Color-coded for integration with HACCP and 5S hygienic zoning programs
  • Compliant with FDA and EU food contact regulations
  • Withstands sterilization up to 249.8 °F (121 °C)
  • Operational temperature range: -4 °F to 338 °F (-20 °C to 170 °C)
Catalog No. 50-005-07571 Supplier MSC Supplier No. 80171697
This stiff table and floor scraper features a durable, solid stainless steel (AISI 304) blade designed to effectively remove stubborn food debris, residue, and labels from various surfaces. Purpose-built for food manufacturing, commercial kitchens, and food service environments, its robust construction ensures long-lasting performance in hygienic and high-safety applications.
  • Durable, solid stainless steel blade
  • Compatible with all Vikan Euro threaded handles for extended reach
  • Lightweight design to reduce user fatigue
  • Color-coded for integration with HACCP and 5S hygienic zoning programs
  • Compliant with FDA and EU food contact regulations
  • Withstands sterilization up to 249.8 °F (121 °C)
  • Operational temperature range: -4 °F to 338 °F (-20 °C to 170 °C)