Learn More
New Jersey Fluid Systems LLC PTFE Hose Assembly, Excellent Chemical Compatibility, High Pressure Resistance, 1 in
Catalog No. NC3763016
Encompass
Introducing the high-performance PTFE Hose Assembly designed for demanding applications. This durable assembly features a virgin PTFE core with dual 304 stainless steel braid reinforcement, ensuring excellent chemical compatibility and superior pressure resistance. With an operating temperature range of -65 to 400°F and a maximum working pressure of 1000 psig, it stands out for critical fluid transfer systems, making it the optimal choice for industries requiring reliability and flexibility.
Catalog No. NC3763016 Supplier New Jersey Fluid Systems LLC Supplier No. SSST16PM16PM16120
Introducing the high-performance PTFE Hose Assembly designed for demanding applications. This durable assembly features a virgin PTFE core with dual 304 stainless steel braid reinforcement, ensuring excellent chemical compatibility and superior pressure resistance. With an operating temperature range of -65 to 400°F and a maximum working pressure of 1000 psig, it stands out for critical fluid transfer systems, making it the optimal choice for industries requiring reliability and flexibility.