Learn More
New Jersey Fluid Systems LLC SS TUBE FITTING MALE 1/4"X3/8"
Catalog No. NC0660053
Encompass
316 Stainless Steel Swagelok Tube Adapter Fitting. 1/4 in. OD Tube Stub x 3/8 in. Male NPT
Catalog No. NC0660053 Supplier New Jersey Fluid Systems LLC Supplier No. SS4TA16
316 Stainless Steel Swagelok Tube Adapter Fitting. 1/4 in. OD Tube Stub x 3/8 in. Male NPT