Learn More
Perkin Elmer US LLC Stainless Steel ATD Prepacked Sample Tubes, Sorbent: Tenax™ TA60/80 Pkg 10
Catalog No. 509052372
Encompass_Preferred
The Tenax TA is conditioned at 320 °C. PerkinElmer recommends a maximum temperature of 350 °C for this sample tube with a desorption temperature of 300 °C. The tubes approximate analyte volatility range is n-C7 to n-C26 and the typical analytes for the Tenax TA are aromatics except benzene, apolar compounds (BP>100 °C) and less volatile compounds.
Catalog No. 50-905-2372 Supplier Perkin Elmer US LLC Supplier No. N9307005
The Tenax TA is conditioned at 320 °C. PerkinElmer recommends a maximum temperature of 350 °C for this sample tube with a desorption temperature of 300 °C. The tubes approximate analyte volatility range is n-C7 to n-C26 and the typical analytes for the Tenax TA are aromatics except benzene, apolar compounds (BP>100 °C) and less volatile compounds.