Learn More
Analytical Sales & Services SS Tubing 1/16" OD - 0.005" ID x 25', Red
Catalog No. 5000806670
Encompass
Pre-cut Stainless Steel Tubing 1/16" OD - 0.005" ID x 25', Red
Catalog No. 50-008-06670 Supplier Analytical Sales & Services Supplier No. 64160
Pre-cut Stainless Steel Tubing 1/16" OD - 0.005" ID x 25', Red