Learn More
Analytical Sales & Services SS Tubing 1/16" OD - 0.007" ID x 50cm, Black
Catalog No. 5000806478
Encompass
Pre-cut Stainless Steel Tubing 1/16" OD - .007" ID x 50cm, Black
Catalog No. 50-008-06478 Supplier Analytical Sales & Services Supplier No. 64130
Pre-cut Stainless Steel Tubing 1/16" OD - .007" ID x 50cm, Black